Dysprosium trifluoride, anhydrous ≥99.9% (REO, rare earth oxide basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
41409-09
41409-18
CAAA41409-09EA
243.32
CAD
CAAA41409-09
CAAA41409-18
Dysprosium trifluoride, anhydrous ≥99.9% (REO, rare earth oxide basis)
Dysprosium trifluoride
For vacuum deposition
Purity: 99.9%
CAS No.: 13569-80-7
Molecular Formula: DyF3
Formula Weight: 219.50
Physical Form: 12mm & down Pieces
Density: 7.5
Appearance: White
Odor: Odorless
Melting Point: 1360°
Boiling Point: 2200°
MDL No.: MFCD00016071
For vacuum deposition
Formula:
DyF₃ MW: 219.5 g/mol Boiling Pt: 2200 °C Melting Pt: 1360 °C Density: 7.5 Storage Temperature: Ambient |
MDL Number:
MFCD00016071 CAS Number: 13569-80-7 EINECS: 236-992-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...