Cerium trifluoride, anhydrous ≥99.9% (REO, rare earth oxide basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
41408-09
41408-18
CAAA41408-09EA
127.03
CAD
CAAA41408-09
CAAA41408-18
Cerium trifluoride, anhydrous ≥99.9% (REO, rare earth oxide basis)
Cerium trifluoride
For vacuum deposition
Purity: 99.9%
CAS No.: 7758-88-5
Molecular Formula: CeF3
Formula Weight: 197.12
Physical Form: 3-6mm Pieces
Density: 6.16
Melting Point: 1460°
Boiling Point: ca 2300°
Merck Reference: 14,1998
MDL No.: MFCD00010932
For vacuum deposition
Formula:
CeF₃ Boiling Pt: ∼2300 °C Melting Pt: 1460 °C Density: 6.16 Storage Temperature: Ambient |
MDL Number:
MFCD00010932 CAS Number: 7758-88-5 EINECS: 231-841-3 Merck Index: 14,1998 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...