Chloroform-[D₁] (100% D) (Isotopic) + 0.03% v/v TMS for NMR spectroscopy
Supplier: Thermo Scientific Chemicals
|
Warning
|
41388-KU
41388-KV
CAAA41388-KUEA
130.82
CAD
CAAA41388-KU
CAAA41388-KV
Chloroform-[D₁] (100% D) (Isotopic) + 0.03% v/v TMS for NMR spectroscopy
Chloroform-[D1]
MDL: MFCD00000827
Notes: Minimum isotopic purity 99.96%
Purity: 100%
CAS No.: 865-49-6
Molecular Formula: CDCl3
Formula Weight: 120.38
Storage Temperature: 2°C to 8°C
Physical Form: Liquid in prescored ampoules, 0.75ml/ampoule
Density: 1.50
Refractive Index: 1.4445
Sensitivity: Moisture and Light Sensitive
Appearance: Colorless
Melting Point: -64°
Boiling Point: 62°
UN No.: 1888
MDL No.: MFCD00000827
Formula:
CDCl₃ Boiling Pt: 62 °C Melting Pt: –64 °C Density: 1.50 Storage Temperature: Refrigerator |
MDL Number:
MFCD00000827 CAS Number: 865-49-6 EINECS: 212-742-4 UN: 1888 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...