Xylenol orange tetrasodium salt ACS

Supplier: Thermo Scientific Chemicals

Synonyms: Xylenol orange water soluble, 2,2',2'',2'''-((((1,1-Dioxido-3H-benzo[c][1,2]oxathiole-3,3-diyl)bis(6-hydroxy-5-methyl-3,1-phenylene))bis(methylene))bis(azanetriyl))tetraacetic acid, sodium salt (1:4)

41379-04 41379-09 41379-18
CAAA41379-04EA 87.53 CAD
CAAA41379-04 CAAA41379-09 CAAA41379-18
Xylenol orange tetrasodium salt ACS
Xylenol orange tetrasodium salt

MDL: MFCD00044293


CAS No.: 3618-43-7 Molecular Formula: C31H28N2Na4O13S Formula Weight: 760.59 Physical Form: Powder Odor: Odorless Melting Point: 210° dec. MDL No.: MFCD00044293

Formula: C₃₁H₂₈N₂Na₄O₁₃S
MW: 760.59 g/mol
Melting Pt: 210 (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00044293
CAS Number: 3618-43-7
EINECS: 222-805-8

Order Now

Specification Test Results

Clarity of solution Pass test
Suitability for zinc titration Pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR