Lead ≥99.999% (metals basis), granules 0.6-4.0 mm, (0.02-0.12 in)

Supplier: Thermo Scientific Chemicals
Danger

41370-14 41370-22 41370-36
CAAA41370-14EA 152.79 CAD
CAAA41370-14 CAAA41370-22 CAAA41370-36
Lead ≥99.999% (metals basis), granules 0.6-4.0 mm, (0.02-0.12 in)
Lead
Formula: Pb
Boiling Pt: 1740 °C (1013 hPa)
Melting Pt: 327 °C
Density: 11.3437 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134050
CAS Number: 7439-92-1
EINECS: 231-100-4
Merck Index: 12,05410

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR