Aluminium ≥99.0% (metals basis), thin foil, Thickness: 8.0 micron (0.0003 in)
Supplier: Thermo Scientific Chemicals
Synonyms:
Aluminum
41358-FI
41358-GH
CAAA41358-FIEA
219.62
CAD
CAAA41358-FI
CAAA41358-GH
Aluminium ≥99.0% (metals basis), thin foil, Thickness: 8.0 micron (0.0003 in)
Aluminium
MDL: MFCD00134029
Purity: 99.0%
CAS No.: 7429-90-5
Physical Form: ≈0.054g/50x50mm, not certified pinhole free
Odor: Odorless
MDL No.: MFCD00134029
Formula:
Al MW: 26.98 g/mol Boiling Pt: 2327 Melting Pt: 660 Density: 2.7 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134029 CAS Number: 7429-90-5 EINECS: 231-072-3 Merck Index: 13,00321 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...