Allyl acetate 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Acetic acid allyl ester
41353-AD
41353-AK
CAAA41353-ADEA
52.46
CAD
CAAA41353-AD
CAAA41353-AK
Allyl acetate 99%
Allyl acetate
MDL: MFCD00008721
Purity: 99%
CAS No.: 591-87-7
Molecular Formula: CH3CO2CH2CH=CH2
Formula Weight: 100.12
Physical Form: Liquid
Density: 0.928
Refractive Index: 1.4042
Melting Point: 6°
Boiling Point: 103-104°
Merck Reference: 14,285
UN No.: 2333
MDL No.: MFCD00008721
Formula:
C₅H₈O₂ MW: 100.12 g/mol Boiling Pt: 103 Melting Pt: -96 Density: 0.928 Flash Pt: 6 Storage Temperature: Ambient |
MDL Number:
MFCD00008721 CAS Number: 591-87-7 EINECS: 209-734-8 UN: 2333 ADR: 3 II Merck Index: 14,285 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...