Chlorodiphenylphosphine 98+%, rods
Supplier: Thermo Scientific Chemicals
|
Danger
|
41352-09
41352-18
41352-30
CAAA41352-09EA
49.14
CAD
CAAA41352-09
CAAA41352-18
CAAA41352-30
Chlorodiphenylphosphine 98+%, rods
Chlorodiphenylphosphine
MDL: MFCD00000529
Beilstein Registry No.: 512032
Fieser: 11,120 18,99
Purity: 98+%
CAS No.: 1079-66-9
Molecular Formula: C12H10ClP
Formula Weight: 220.64
Flash Point: 138°(280°F)
Physical Form: Liquid
Density: 1.204
Refractive Index: 1.6360
Sensitivity: Air and Moisture Sensitive
Appearance: Colorless to pale yellow
Odor: Acrid
Melting Point: 14-16°
Boiling Point: 318-321°
Beilstein Reference: 512032
UN No.: 3265
MDL No.: MFCD00000529
Formula:
C₁₂H₁₀ClP Boiling Pt: 318…321 °C Melting Pt: 14…16 °C Density: 1,204 Flash Pt: 138 °C (280 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000529 CAS Number: 1079-66-9 EINECS: 214-093-2 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...