Benzalkonium chloride

Supplier: Thermo Scientific Chemicals
Danger

41339-14 41339-22 41339-36
CAAA41339-14EA 45.31 CAD
CAAA41339-14 CAAA41339-22 CAAA41339-36
Benzalkonium chloride
Benzalkonium chloride

Cationic surface active agent


CAS No.: 63449-41-2 Molecular Formula: C6H5CH2N(CH3)2RCl Physical Form: Semi-solid/solid, R=C8H17 to C18H37 Solubility: Soluble in water Merck Reference: 14,1059 UN No.: 2923 MDL No.: MFCD00145757


Cationic surface active agent

Formula: C₆H₅CH₂N(CH₃)₂RCl (R=C₈H₁₇ to C₁₈H₃₇)
Melting Pt: 29…34 °C
Density: 0.98 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00145757
CAS Number: 63449-41-2
EINECS: 264-151-6
UN: 2923
ADR: 8,III
Merck Index: 14,1059

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR