8-Quinolinol ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
8-Hydroxyquinoline
A bacteriostatic and fungistatic agent. Also a chelating agent for metals
CAS No.: 148-24-3
Molecular Formula: C9H7NO
Formula Weight: 145.16
Physical Form: Granular
Sensitivity: Light Sensitive
Appearance: White to pale yellow
Melting Point: 72-76°
Boiling Point: 267°
Solubility: Almost insoluble in water and diethyl ether. Highly soluble in ethanol, acetone, chloroform, and benzene.
Beilstein Reference: 114512
Merck Reference: 14,4843
MDL No.: MFCD00006807
A bacteriostatic and fungistatic agent. Also a chelating agent for metals
Formula:
C₉H₇NO Boiling Pt: 267 °C Melting Pt: 72…76 °C Density: 1.03 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006807 CAS Number: 148-24-3 EINECS: 205-711-1 Merck Index: 14,4843 |
|
Specification Test Results
Insoluble in alcohol | 0.05% |
Melting point | 72.5-74.0° |
Residue after ignition | 0.05% |
SO3 | 0.0002 |
Suitability for magnesium determination | Pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...