Benzyl alcohol 99+% ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
41218-M1
41218-AK
41218-K2
41218-K7
CAAA-41218-M1EA
692.04
CAD
CAAA-41218-M1
CAAA41218-AK
CAAA41218-K2
CAAA41218-K7
Benzyl alcohol 99+% ACS
Benzyl alcohol
MDL: MFCD00004599
Beilstein Registry No.: 878307
Purity: 99+%
CAS No.: 100-51-6
Molecular Formula: C7H8O
Formula Weight: 108.14
Flash Point: 100°(212°F)
Physical Form: Liquid
Density: 1.045
Refractive Index: 1.5400
Odor: Aromatic
Melting Point: -15°
Boiling Point: 205°
Beilstein Reference: 878307
Merck Reference: 14,1124
MDL No.: MFCD00004599
Formula:
C₆H₅CH₂OH Boiling Pt: 205 °C Melting Pt: –15 °C Density: 1.045 Flash Pt: 100 °C (212 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004599 CAS Number: 100-51-6 EINECS: 202-859-9 Merck Index: 14,1124 |
|
Specification Test Results
Color (APHA) | 20 |
Residue after ignition | 0.005% |
C3H5COCH3 | 0.02% |
C3H5CHO | 0.01% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...