Dipotassium hexahydroxoplatinate ≥99.95% (metals basis) Pt ≥ 51.5%, Premion®
Supplier: Thermo Scientific Chemicals
|
Warning
|
41014-03
41014-06
CAAA41014-03EA
312.84
CAD
CAAA41014-03
CAAA41014-06
Dipotassium hexahydroxoplatinate ≥99.95% (metals basis) Pt ≥ 51.5%, Premion®
Dipotassium hexahydroxoplatinate
MDL: MFCD00064671
Purity: 99.95%
CAS No.: 12285-90-4
Molecular Formula: K2Pt(OH)6
Formula Weight: 375.34
Physical Form: Powder
Formula:
H₆K₂O₆Pt Storage Temperature: Ambient |
MDL Number:
MFCD00064671 CAS Number: 12285-90-4 EINECS: 235-554-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...