Petroleum spirit, 35…60 °C
Supplier: Thermo Scientific Chemicals
|
Danger
|
40982-K2
40982-K7
CAAA40982-K2EA
78.37
CAD
CAAA40982-K2
CAAA40982-K7
Petroleum spirit, 35…60 °C
Petroleum spirit
Suitable for environmental and pesticide analysis
CAS No.: 8032-32-4
Flash Point: -49°(-57°F)
Physical Form: Liquid
Density: 0.640
Refractive Index: 1.3630
Appearance: Colorless
Odor: Petroleum-like
Boiling Point: 35-60°
UN No.: 1268
MDL No.: MFCD00081849
Suitable for environmental and pesticide analysis
Boiling Pt:
35…60 °C Density: 0.640 Flash Pt: –49 °C (–57 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00081849 CAS Number: 64742-49-0 EINECS: 232-453-7 UN: 1268 ADR: 3,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...