Diethyl ether 99+% inhibitor-free for spectrophotometry Inhibitor-free
Supplier: Thermo Scientific Chemicals
|
Danger
|
40976-K2
40976-M1
CAAA40976-K2EA
146.29
CAD
CAAA40976-K2
CAAA40976-M1
Diethyl ether 99+% inhibitor-free for spectrophotometry Inhibitor-free
Diethyl ether
Solvent for U.V. spectrophotometric and general use
Purity: 99+%
CAS No.: 60-29-7
Molecular Formula: C4H10O
Formula Weight: 74.12
Flash Point: -45°(-49°F)
Physical Form: Liquid
Density: 0.713
Refractive Index: 1.3525
Appearance: Colorless
Odor: Sweet
Melting Point: -116°
Boiling Point: 34.6°
Merck Reference: 14,3806
UN No.: 1155
MDL No.: MFCD00011646
Solvent for U.V. spectrophotometric and general use
Formula:
(CH₃CH₂)₂O Boiling Pt: 34.6 °C Melting Pt: –116 °C Density: 0.713 Flash Pt: –45 °C (–49 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00011646 CAS Number: 60-29-7 EINECS: 200-467-2 UN: 1155 ADR: 3,I REACH: 01-2119535785-29 Merck Index: 14,3806 |
|
Specification Test Results
Evaporation residue | 5 ppm |
Peroxide (as H2O2) | 5ppm ppm |
H2O | 0.05% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...