Cyclohexane 99,7, Environmental Grade

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Hexahydrobenzene, Hexamethylene

40975-K7 40975-M6
CAAA40975-K7EA 265.44 CAD
CAAA40975-K7 CAAA40975-M6
Cyclohexane 99,7, Environmental Grade
Cyclohexane

Suitable for pesticide residue, environmental and GC analysis


Purity: 99.7+% CAS No.: 110-82-7 Molecular Formula: C6H12 Formula Weight: 84.16 Flash Point: -18°(-1°F) Physical Form: Liquid Density: 0.779 Refractive Index: 1.4260 Appearance: Colorless Odor: Pungent Melting Point: 6.5° Boiling Point: 80-81° Beilstein Reference: 1900225 Merck Reference: 14,2723 UN No.: 1145 MDL No.: MFCD00003814


Suitable for pesticide residue, environmental and GC analysis

Formula: C₆H₁₂
MW: 84.16 g/mol
Boiling Pt: 81
Melting Pt: 7
Density: 0.779
Flash Pt: –18 °C (–1 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003814
CAS Number: 110-82-7
EINECS: 203-806-2
UN: 1145
ADR: 3 II
Merck Index: 14,2723
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a1e51ba998bef126804c3fd25b361be72cd9545&vid=d1e7f75d9f2a944eb15a7740c5254588cc9a4e67&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR