Platinum iridium alloy (90:10 w%), gauze woven from wire 150 mesh, Ø 0.043 mm (0.0017 in)
Supplier: Thermo Scientific Chemicals
40934-FF
40934-FI
CAAA40934-FFEA
510.34
CAD
CAAA40934-FF
CAAA40934-FI
Platinum iridium alloy (90:10 w%), gauze woven from wire 150 mesh, Ø 0.043 mm (0.0017 in)
Platinum Iridium alloy (90:10 w%)
Applications: Filtration, electrodes
Molecular Formula: Pt:Ir; 90:10 wt%
Physical Form: Wire Cloth
Odor: Odorless
MDL No.: MFCD02091683
Filtration, electrodes
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...