Tungsten, gauze woven from wire 20 mesh, Ø 0.127 mm (0.005 in)

Supplier: Thermo Scientific Chemicals

40922-FL 40922-GT 40922-HB
CAAA40922-FLEA 225.94 CAD
CAAA40922-FL CAAA40922-GT CAAA40922-HB
Tungsten, gauze woven from wire 20 mesh, Ø 0.127 mm (0.005 in)
Tungsten

Filtration, electrodes


CAS No.: 7440-33-7 Physical Form: Wire Cloth MDL No.: MFCD00011461


Filtration, electrodes

Formula: W
Boiling Pt: 5660 °C (1013 hPa)
Melting Pt: 3410 °C
Density: 19 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011461
CAS Number: 7440-33-7
EINECS: 231-143-9
Merck Index: 12,09945

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR