Titanium, gauze woven from wire 50 mesh, Ø 0.102 mm (0.004 in)

Supplier: Thermo Scientific Chemicals
Warning

40921-FL 40921-GT 40921-HB
CAAA40921-FLEA 417.12 CAD
CAAA40921-FL CAAA40921-GT CAAA40921-HB
Titanium, gauze woven from wire 50 mesh, Ø 0.102 mm (0.004 in)
Titanium

MDL: MFCD00011264 Notes: Open area: 64%; Width of opening: 0.016in


CAS No.: 7440-32-6 Physical Form: Wire Cloth Odor: Odorless MDL No.: MFCD00011264

Formula: Ti
Boiling Pt: 3262 °C (1013 hPa)
Melting Pt: 1660 °C
Density: 4.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR