Titanium, gauze woven from wire 30 mesh, Ø 0.102 mm (0.004 in)
Supplier: Thermo Scientific Chemicals
|
Warning
|
40919-FL
40919-GT
40919-HB
CAAA40919-FLEA
233.84
CAD
CAAA40919-FL
CAAA40919-GT
CAAA40919-HB
Titanium, gauze woven from wire 30 mesh, Ø 0.102 mm (0.004 in)
Titanium
MDL: MFCD00011264
Notes: Open area: 77.4%; Width of opening: 0.029in
CAS No.: 7440-32-6
Physical Form: Wire Cloth
Odor: Odorless
MDL No.: MFCD00011264
Formula:
Ti Boiling Pt: 3262 °C (1013 hPa) Melting Pt: 1660 °C Density: 4.51 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011264 CAS Number: 7440-32-6 EINECS: 231-142-3 Merck Index: 12,09610 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...