Titanium, gauze woven from wire 20 mesh, Ø 0.076 mm (0.003 in)

Supplier: Thermo Scientific Chemicals
Warning

40918-FL 40918-HB
CAAA40918-FLEA 161.16 CAD
CAAA40918-FL CAAA40918-GT CAAA40918-HB
Titanium, gauze woven from wire 20 mesh, Ø 0.076 mm (0.003 in)
Titanium

MDL: MFCD00011264 Notes: Open area: 88.4%; Width of opening: 0.047in


CAS No.: 7440-32-6 Physical Form: Wire Cloth Odor: Odorless MDL No.: MFCD00011264

Formula: Ti
MW: 47.88 g/mol
Boiling Pt: 3262
Melting Pt: 1660
Density: 4.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR