Molybdenum, gauze woven from wire 100 mesh, Ø 0.0636 mm (0.0025 in)

Supplier: Thermo Scientific Chemicals

40912-FL 40912-GT 40912-HB
CAAA40912-FLEA 260.7 CAD
CAAA40912-FL CAAA40912-GT CAAA40912-HB
Molybdenum, gauze woven from wire 100 mesh, Ø 0.0636 mm (0.0025 in)
Molybdenum

MDL: MFCD00003465 Notes: Open area: 56.3%; Width of opening: 0.0075in


CAS No.: 7439-98-7 Physical Form: Wire Cloth MDL No.: MFCD00003465

Formula: Mo
Boiling Pt: 3617 °C (1013 hPa)
Melting Pt: 2622 °C
Density: 10.28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003465
CAS Number: 7439-98-7
EINECS: 231-107-2
Merck Index: 12,06317

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR