Molybdenum, gauze woven from wire 50 mesh, Ø 0.0509 mm (0.002 in)

Supplier: Thermo Scientific Chemicals

40911-FL 40911-GT 40911-HB
CAAA40911-FLEA 152 CAD
CAAA40911-FL CAAA40911-GT CAAA40911-HB
Molybdenum, gauze woven from wire 50 mesh, Ø 0.0509 mm (0.002 in)
Molybdenum

MDL: MFCD00003465 Notes: Open area: 81%; Width of opening: 0.018in


CAS No.: 7439-98-7 Physical Form: Wire Cloth MDL No.: MFCD00003465

Formula: Mo
Boiling Pt: 3617 °C (1013 hPa)
Melting Pt: 2622 °C
Density: 10.28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003465
CAS Number: 7439-98-7
EINECS: 231-107-2
Merck Index: 12,06317

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR