Molybdenum, gauze woven from wire 20 mesh, Ø 0.305 mm (0.012 in)

Supplier: Thermo Scientific Chemicals

40908-FL 40908-GT 40908-HB
CAAA40908-FLEA 97.64 CAD
CAAA40908-FL CAAA40908-GT CAAA40908-HB
Molybdenum, gauze woven from wire 20 mesh, Ø 0.305 mm (0.012 in)
Molybdenum

Filtration, electrodes


CAS No.: 7439-98-7 Physical Form: Wire Cloth MDL No.: MFCD00003465


Filtration, electrodes

Formula: Mo
Boiling Pt: 3617 °C (1013 hPa)
Melting Pt: 2622 °C
Density: 10.28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003465
CAS Number: 7439-98-7
EINECS: 231-107-2
Merck Index: 12,06317

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR