Copper, gauze woven from wire 40 mesh, Ø 0.056 mm (0.0022 in)
Supplier: Thermo Scientific Chemicals
40907-FL
40907-GT
40907-HB
CAAA40907-FLEA
104.75
CAD
CAAA40907-FL
CAAA40907-GT
CAAA40907-HB
Copper, gauze woven from wire 40 mesh, Ø 0.056 mm (0.0022 in)
Copper
MDL: MFCD00010965
Notes: Open area: 83.2%; Width of opening: 0.0228in
CAS No.: 7440-50-8
Physical Form: Wire Cloth
Odor: Odorless
MDL No.: MFCD00010965
Formula:
Cu Boiling Pt: 2595 °C (1013 hPa) Melting Pt: 1083 °C Density: 8.94 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010965 CAS Number: 7440-50-8 EINECS: 231-159-6 Merck Index: 13,02545 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...