Aluminium ≥99.999% (metals basis), sputtering target, Ø 76.2 mm (3.0 in), Thickness: 6.35 mm (0.250 in)

Supplier: Thermo Scientific Chemicals

Synonyms: Aluminum

40829-KS 40829-KS
CAAA40829-KSEA 676.24 CAD
CAAA40829-KS AA40829-KS
Aluminium ≥99.999% (metals basis), sputtering target, Ø 76.2 mm (3.0 in), Thickness: 6.35 mm (0.250 in)
Aluminium
Formula: Al
MW: 26.98 g/mol
Boiling Pt: 2327 °C (1013 hPa)
Melting Pt: 660 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
Merck Index: 13,00321

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR