Polypropylene glycol, M.W. ∼4,000

Supplier: Thermo Scientific Chemicals

Synonyms: Poly(propylene oxide), PPG

40815-18 40815-30
CAAA40815-18EA 56.41 CAD
CAAA40815-18 CAAA40815-30
Polypropylene glycol, M.W. ∼4,000
Polypropylene glycol

MDL: MFCD01779690 Notes: Average Mí 4000


CAS No.: 25322-69-4 Molecular Formula: H[OCH(CH3)CH2]nOH Physical Form: Liquid MDL No.: MFCD01779690

Formula: H(OCH₂CH₂CH₂)nOH
Storage Temperature: Ambient
MDL Number: MFCD00084446
CAS Number: 25322-69-4
EINECS: 500-039-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR