Polypropylene glycol, M.W. ∼ 3,000
Supplier: Thermo Scientific Chemicals
Synonyms:
Poly(propylene oxide), PPG
40814-18
40814-30
CAAA40814-18EA
47.56
CAD
CAAA40814-18
CAAA40814-30
Polypropylene glycol, M.W. ∼ 3,000
Polypropylene glycol
MDL: MFCD00084446
Notes: Average Mí 3000
CAS No.: 25322-69-4
Molecular Formula: H[OCH(CH3)CH2]nOH
Physical Form: Liquid
MDL No.: MFCD00084446
Formula:
H(OCH₂CH₂CH₂)nOH Storage Temperature: Ambient |
MDL Number:
MFCD00084446 CAS Number: 25322-69-4 EINECS: 500-039-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...