Polypropylene glycol, M.W. 1,000

Supplier: Thermo Scientific Chemicals

40812-18 40812-30
CAAA40812-18EA 43.61 CAD
CAAA40812-18 CAAA40812-30
Polypropylene glycol, M.W. 1,000
Polypropylene glycol

MDL: MFCD01779683 Notes: Average Mí 1000


CAS No.: 25322-69-4 Molecular Formula: H[OCH(CH3)CH2]nOH Physical Form: Liquid Appearance: Colorless MDL No.: MFCD01779683

Formula: H(OCH₂CH₂CH₂)nOH
Storage Temperature: Ambient
MDL Number: MFCD00084446
CAS Number: 25322-69-4
EINECS: 500-039-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR