Polypropylene glycol, M.W. 400

Supplier: Thermo Scientific Chemicals

Synonyms: Poly(propylene oxide), PPG

40811-18 40811-30
CAAA40811-18EA 47.24 CAD
CAAA40811-18 CAAA40811-30
Polypropylene glycol, M.W. 400
Polypropylene glycol

MDL: MFCD01779680 Notes: Average Mí 400


CAS No.: 25322-69-4 Molecular Formula: H[OCH(CH3)CH2]nOH Physical Form: Liquid MDL No.: MFCD01779680

Formula: H(OCH₂CH₂CH₂)nOH
Storage Temperature: Ambient
MDL Number: MFCD00084446
CAS Number: 25322-69-4
EINECS: 500-039-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR