Manganese(II) carbonate ≥99.985% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals

40756-06 40756-14 40756-22
CAAA40756-06EA 70.78 CAD
CAAA40756-06 CAAA40756-14 CAAA40756-22
Manganese(II) carbonate ≥99.985% (metals basis), Puratronic®
Manganese (II) carbonate

MDL: MFCD00011116


Purity: 99.985% CAS No.: 598-62-9 Molecular Formula: CMnO3 Formula Weight: 114.95 Physical Form: Powder Density: 3.125 Appearance: Pale brown to dark purple Odor: Odorless Melting Point: 350° dec. Merck Reference: 14,5726 MDL No.: MFCD00011116

Formula: CMnO₃
Melting Pt: 350 °C (dec.)
Density: 3.125
Storage Temperature: Ambient
MDL Number: MFCD00011116
CAS Number: 598-62-9
EINECS: 209-942-9
Merck Index: 14,5726

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR