Magnesium fluoride ≥99.99% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Warning
|
40755-04
40755-09
40755-18
CAAA40755-04EA
113.6
CAD
CAAA40755-04
CAAA40755-09
CAAA40755-18
Magnesium fluoride ≥99.99% (metals basis), Puratronic®
Magnesium fluoride
MDL: MFCD00011108
Purity: 99.99%
CAS No.: 7783-40-6
Molecular Formula: F2Mg
Formula Weight: 62.30
Physical Form: Powder
Density: 3.148
Refractive Index: 1.35-1.38
Appearance: White
Odor: Odorless
Melting Point: 1261°
Merck Reference: 14,5665
MDL No.: MFCD00011108
Formula:
MgF₂ Boiling Pt: 2260 °C (1013 hPa) Melting Pt: 1261 °C Density: 3.148 Storage Temperature: Ambient |
MDL Number:
MFCD00011108 CAS Number: 7783-40-6 EINECS: 231-995-1 Merck Index: 14,5665 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...