Copper(I) bromide ≥99.998% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Warning

40752-03 40752-06
CAAA40752-03EA 149.15 CAD
CAAA40752-03 CAAA40752-06
Copper(I) bromide ≥99.998% (metals basis), Puratronic®
Copper (I) bromide

MDL: MFCD00010969


Purity: 99.998% CAS No.: 7787-70-4 Molecular Formula: CuBr Formula Weight: 143.45 Physical Form: Powder, Ampouled under argon Density: 4.98 Appearance: Pale green Melting Point: 492° Boiling Point: 1345° Merck Reference: 14,2659 UN No.: 3077 MDL No.: MFCD00010969

Formula: BrCu
MW: 143.45 g/mol
Boiling Pt: 1345 °C
Melting Pt: 492 °C
Density: 4.98
Storage Temperature: Ambient
MDL Number: MFCD00010969
CAS Number: 7787-70-4
EINECS: 232-131-6
UN: 3077
ADR: 9,III
Merck Index: 14,2659

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR