Ammonium iron (II) sulfate hexahydrate ≥99.999% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Warning

40751-03 40751-06 40751-14
CAAA40751-03EA 36.18 CAD
CAAA40751-03 CAAA40751-06 CAAA40751-14
Ammonium iron (II) sulfate hexahydrate ≥99.999% (metals basis), Puratronic®
Ammonium iron (II) sulphate hexahydrate

MDL: MFCD00150530


Purity: 99.999% CAS No.: 7783-85-9 Molecular Formula: H8FeN2O8S2.xH2O (x≈6) Formula Weight: 284.04(anhy) Physical Form: Coarse Powder Density: 1.864 Appearance: Pale bluish green Odor: Odorless Melting Point: ca 100° dec. Merck Reference: 14,521 MDL No.: MFCD00150530

Formula: (NH₄)₂Fe(SO₄)₂·6H₂O
Melting Pt: ∼ 100 °C (dec.)
Density: 1.864
Storage Temperature: Ambient
MDL Number: MFCD00150530
CAS Number: 7783-85-9
EINECS: 233-382-4
Merck Index: 14,521

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR