Silver ≥99.95% (metals basis), thin foil, Thickness: 0.0125 mm (0.0005 in)

Supplier: Thermo Scientific Chemicals

40749-FF 40749-FI 40749-GH
CAAA40749-FFEA 409.22 CAD
CAAA40749-FF CAAA40749-FI CAAA40749-GH
Silver ≥99.95% (metals basis), thin foil, Thickness: 0.0125 mm (0.0005 in)
Silver

MDL: MFCD00003397


Purity: 99.95+% CAS No.: 7440-22-4 Physical Form: ≈0.08g/25x25mm, Not certified pinhole free Odor: Characteristic MDL No.: MFCD00003397

Formula: Ag
MW: 107.87 g/mol
Boiling Pt: 2212
Melting Pt: 962
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR