Palladium ≥99.95% (metals basis), thin foil, Thickness: 0,0125 mm (0.0005 in)

Supplier: Thermo Scientific Chemicals

40744-FF 40744-FI
CAAA40744-FFEA 494.54 CAD
CAAA40744-FF CAAA40744-FI
Palladium ≥99.95% (metals basis), thin foil, Thickness: 0,0125 mm (0.0005 in)
Palladium

MDL: MFCD00011167


Purity: 99.95% CAS No.: 7440-05-3 Physical Form: ≈0.094g/25x25mm, not certified pinhole free Odor: Odorless MDL No.: MFCD00011167

Formula: Pd
Boiling Pt: 3167 °C (1013 hPa)
Melting Pt: 1555 °C
Density: 12.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6
Merck Index: 12,07121

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR