Tantalum ≥99.9% (metals basis), thin foil, thickness 0.005 mm (0.0002 in)
Supplier: Thermo Scientific Chemicals
40717-FI
40717-FF
40717-FF
40717-FI
CAAA40717-FIEA
641.01
CAD
AA40717-FF
CAAA40717-FI
AA40717-FI
CAAA40717-FF
Tantalum ≥99.9% (metals basis), thin foil, thickness 0.005 mm (0.0002 in)
Tantalum
Formula:
Ta Boiling Pt: 5427 °C (1013 hPa) Melting Pt: 2996 °C Density: 16,69 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011252 CAS Number: 7440-25-7 Merck Index: 12,09223 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...