Tantalum ≥99.9% (metals basis), thin foil, Thickness: 0.0025 mm (0.0001 in)

Supplier: Thermo Scientific Chemicals

40716-FF 40716-FI
CAAA40716-FFEA 657.28 CAD
CAAA40716-FF CAAA40716-FI CAAA40716-GH
Tantalum ≥99.9% (metals basis), thin foil, Thickness: 0.0025 mm (0.0001 in)
Tantalum

MDL: MFCD00011252


Purity: 99.9% CAS No.: 7440-25-7 Physical Form: ≈0.026g/25x25mm, not certified pinhole free Odor: Odorless MDL No.: MFCD00011252

Formula: Ta
Boiling Pt: 5427 °C (1013 hPa)
Melting Pt: 2996 °C
Density: 16.69 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011252
CAS Number: 7440-25-7
EINECS: 231-135-5
Merck Index: 12,09223

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR