Lithium iodide, anhydrous 99,95 Metals basis

Supplier: Thermo Scientific Chemicals
Warning

40666-14 40666-22 40666-36
CAAA40666-14EA 225.94 CAD
CAAA40666-14 CAAA40666-22 CAAA40666-36
Lithium iodide, anhydrous 99,95 Metals basis
Lithium iodide

MDL: MFCD00011092


Purity: 99.95% CAS No.: 10377-51-2 Molecular Formula: ILi Formula Weight: 133.84 Physical Form: Crystalline/Powder, Ampouled under argon Density: 4.076 Refractive Index: 1.955 Sensitivity: Light Sensitive and Hygroscopic Appearance: White Odor: Odorless Melting Point: 449° Boiling Point: 1170-1190° Merck Reference: 14,5535 MDL No.: MFCD00011092

Formula: ILi
MW: 133.85 g/mol
Boiling Pt: 1177
Melting Pt: 450
Density: 4.076
Storage Temperature: Ambient
MDL Number: MFCD00011092
CAS Number: 10377-51-2
EINECS: 233-822-5
Merck Index: 14,5535

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR