Titanium(IV) iodide, anhydrous ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
40663-09
40663-18
CAAA40663-09EA
278.08
CAD
CAAA40663-09
CAAA40663-18
Titanium(IV) iodide, anhydrous ≥99.9% (metals basis)
Titanium (IV) iodide
MDL: MFCD00043077
Purity: 99.9%
CAS No.: 7720-83-4
Molecular Formula: TiI4
Formula Weight: 555.52
Physical Form: Lump, Ampouled under argon
Density: 4.30
Melting Point: 150°
UN No.: 3260
MDL No.: MFCD00043077
Formula:
I₄Ti Boiling Pt: 360 °C (1013 hPa) Melting Pt: 150 °C Density: 4.30 Storage Temperature: Ambient |
MDL Number:
MFCD00043077 CAS Number: 7720-83-4 EINECS: 231-754-0 UN: 3260 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...