Ytterbium(III) chloride, ultra dry ≥99.99% (REO, rare earth oxide basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
40653-06
40653-14
CAAA40653-06EA
249.64
CAD
CAAA40653-06
CAAA40653-14
Ytterbium(III) chloride, ultra dry ≥99.99% (REO, rare earth oxide basis)
Ytterbium (III) chloride
MDL: MFCD00011288
Purity: 99.99%
CAS No.: 10361-91-8
Molecular Formula: YbCl3
Formula Weight: 279.40
Physical Form: -10 Mesh Beads, Ampouled under argon
Sensitivity: Hygroscopic
Merck Reference: 14,10105
MDL No.: MFCD00011288
Formula:
Cl₃Yb Melting Pt: 854 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011288 CAS Number: 10361-91-8 EINECS: 233-800-5 Merck Index: 14,10105 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...