Hafnium dioxide ≥99.99% (metals basis excluding Zr) (max. 100 ppm Zr)

Supplier: Thermo Scientific Chemicals

40620-03 40620-06
CAAA40620-03EA 229.1 CAD
CAAA40620-03 CAAA40620-06
Hafnium dioxide ≥99.99% (metals basis excluding Zr) (max. 100 ppm Zr)
Hafnium dioxide

MDL: MFCD00003565


Purity: 99.99% CAS No.: 12055-23-1 Molecular Formula: HfO2 Formula Weight: 210.49 Physical Form: Powder Density: 9.68 Melting Point: 2774° Merck Reference: 14,4588 MDL No.: MFCD00003565

Formula: HfO₂
MW: 210.49
Boiling Pt: 5400
Melting Pt: 2812
Density: 9.68
Storage Temperature: Ambient
MDL Number: MFCD00003565
CAS Number: 12055-23-1
EINECS: 235-013-2
Merck Index: 14,04727

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR