Antimony(III) potassium oxitartrate trihydrate 99.0-103.0% ACS

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (R,R)-(+)-Antimony potassium tartrate trihydrate, (+)-Antimony potassium L-tartrate trihydrate, Tartar emetic, Antimony potassium L(+)-tartrate trihydrate, L(+)-Tartaric acid antimony potassium salt trihydrate, Potassium Antimony (III) Oxide Tartrate Trihydrate, (R,R)-(+)-Tartaric acid antimony potassium salt trihydrate, L(+)-Antimony potassium tartrate trihydrate, Potassium antimony(III) oxide tartrate trihydrate, Antimony potassium (+) tartrate trihydrate, Potassium antimony tartrate trihydrate, Potassium antimonyl tartrate sesquihydrate, Antimony potassium tartrate trihydrate, Potassium antimonyl tartrate trihydrate

40619-14 40619-22 40619-36
CAAAA40619-14EA 53.09 CAD
CAAAA40619-14 CAAA40619-22 CAAA40619-36
Antimony(III) potassium oxitartrate trihydrate 99.0-103.0% ACS
Antimony (III) potassium oxitartrate trihydrate

MDL: MFCD00148863


Purity: 99.0-103.0% CAS No.: 28300-74-5 Molecular Formula: C8H4K2O12Sb2.3H2O Formula Weight: 667.87 (613.82anhy) Physical Form: Powder Density: 2.6 Appearance: White Odor: Unpleasant Merck Reference: 14,700 UN No.: 1551 MDL No.: MFCD00148863

Formula: C₈H₄K₂O₁₂Sb₂·3H₂O
MW: 667.88 g/mol
Melting Pt: 100 °C
Density: 2.6
Storage Temperature: Ambient
MDL Number: MFCD00167056
CAS Number: 28300-74-5
EINECS: 229-436-1
UN: 1551
ADR: 6.1,III
Merck Index: 14,700

Order Now

Specification Test Results

Form Powder
Loss on drying 0.027
As 0.015%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR