Boron nitride

Supplier: Thermo Scientific Chemicals
Warning

40607-04 40607-09
CAAA40607-04EA 169.06 CAD
CAAA40607-04 CAAA40607-09
Boron nitride
Boron nitride

MDL: MFCD00011317 Notes: Mohs hardness 10. Dissolves in fused sodium carbonate


CAS No.: 10043-11-5 Molecular Formula: BN (cubic) Formula Weight: 24.82 Physical Form: 4-8 Micron Powder Density: 3.48 Odor: Odorless Merck Reference: 14,1346 MDL No.: MFCD00011317

Formula: BN
MW: 24.82 g/mol
Melting Pt: 2527
Density: 3.48
Storage Temperature: Ambient
MDL Number: MFCD00011317
CAS Number: 10043-11-5
EINECS: 233-136-6
Merck Index: 14,1346

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR