Magnesium ≥99.9% (metals basis), foil, hard, Thickness: 1.0 mm (0.04 in)

Supplier: Thermo Scientific Chemicals
Danger

40604-FF 40604-FI 40604-GH
CAAA40604-FFEA 322.32 CAD
CAAA40604-FF CAAA40604-FI CAAA40604-GH
Magnesium ≥99.9% (metals basis), foil, hard, Thickness: 1.0 mm (0.04 in)
Magnesium

MDL: MFCD00085308


Purity: 99.9% CAS No.: 7439-95-4 Physical Form: ≈1.1g/25x25mm, Temper: as rolled Odor: Odorless UN No.: 1869 MDL No.: MFCD00085308

Formula: Mg
Storage Temperature: Ambient
MDL Number: MFCD00085308
CAS Number: 7439-95-4
EINECS: 231-104-6
UN: 1869
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR