Magnesium 99,9 Metals basis, foil, hard, Thickness: 0.25 mm (0.01 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
40603-FF
40603-FI
40603-GH
CAAA40603-FFEA
246.48
CAD
CAAA40603-FF
CAAA40603-FI
CAAA40603-GH
Magnesium 99,9 Metals basis, foil, hard, Thickness: 0.25 mm (0.01 in)
Magnesium
MDL: MFCD00085308
Purity: 99.9%
CAS No.: 7439-95-4
Physical Form: ≈0.27g/25x25mm, Temper: as rolled
Odor: Odorless
UN No.: 1869
MDL No.: MFCD00085308
Formula:
Mg MW: 24.3 g/mol Boiling Pt: 1107 Melting Pt: 651 Storage Temperature: Ambient |
MDL Number:
MFCD00085308 CAS Number: 7439-95-4 EINECS: 231-104-6 UN: 1869 ADR: 4.1 III Merck Index: 13,05675 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...