Gold ≥99.9% (metals basis), gauze woven from wire 82 mesh, Ø 0.06 mm (0.0025 in)

Supplier: Thermo Scientific Chemicals

40586-FF 40586-FI
CAAA40586-FFEA 597.24 CAD
CAAA40586-FF CAAA40586-FI
Gold ≥99.9% (metals basis), gauze woven from wire 82 mesh, Ø 0.06 mm (0.0025 in)
Gold

MDL: MFCD00003436 Notes: Open area: 63.2%; Width of opening: 0.0097in


Purity: 99.9% CAS No.: 7440-57-5 Physical Form: Wire Cloth MDL No.: MFCD00003436

Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR