Cerium(IV) oxide ≥96% (REO, rare earth oxide basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ceric oxide, Cerium dioxide
Polishing and decolorizing glass, in heat-resistant alloy coatings, or infrared filters to prevent reflection, and as a catalyst for organic reactions
Purity: 96%
CAS No.: 1306-38-3
Molecular Formula: CeO2
Formula Weight: 172.12
Physical Form: Powder
Density: 7.132
Appearance: Pale yellow
Odor: Odorless
Melting Point: 2600°
Solubility: Insoluble in water. Soluble in H2SO4 and HNO3
Merck Reference: 14,1989
MDL No.: MFCD00010933
Polishing and decolorizing glass, in heat-resistant alloy coatings, or infrared filters to prevent reflection, and as a catalyst for organic reactions
Formula:
CeO₂ MW: 172.11 g/mol Boiling Pt: 3468 Melting Pt: 2000 Density: 7.132 Storage Temperature: Ambient |
MDL Number:
MFCD00010933 CAS Number: 1306-38-3 EINECS: 215-150-4 Merck Index: 14,1989 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...