Aluminium ≥99.99% (metals basis), foil, annealed, Thickness: 0.5 mm (0.02 in)

Supplier: Thermo Scientific Chemicals

40531-FI 40531-GH 40531-HP
CAAA40531-FIEA 90.69 CAD
CAAA40531-FI CAAA40531-GH CAAA40531-HP
Aluminium ≥99.99% (metals basis), foil, annealed, Thickness: 0.5 mm (0.02 in)
Aluminium

MDL: MFCD00134029


Purity: 99.99% CAS No.: 7429-90-5 Physical Form: ≈3.4g/50x50mm, Temper: soft Odor: Odorless MDL No.: MFCD00134029

Formula: Al
Boiling Pt: 2327 °C (1013 hPa)
Melting Pt: 660 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR