N-Ethylmaleimide (NEM) 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N-Ethylmaleimide, 1-Ethyl-1H-pyrrole-2,5-dione

40526-03 40526-06 40526-14
CAAA40526-03EA 52.61 CAD
CAAA40526-03 CAAA40526-06 CAAA40526-14
N-Ethylmaleimide (NEM) 99+%
N-Ethylmaleimide

MDL: MFCD00005509 Beilstein Registry No.: 112448


Purity: 99+% CAS No.: 128-53-0 Molecular Formula: C6H7NO2 Formula Weight: 125.13 Flash Point: 73°(163°F) Physical Form: Crystalline Sensitivity: Light Sensitive Appearance: White to pale cream Odor: Pungent Melting Point: 43-46° Boiling Point: 209-211° Beilstein Reference: 112448 Merck Reference: 14,3822 UN No.: 2928 MDL No.: MFCD00005509

Formula: C₆H₇NO₂
Boiling Pt: 209…211 °C
Melting Pt: 43…46 °C
Flash Pt: 73 °C (163 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005509
CAS Number: 128-53-0
EINECS: 204-892-4
UN: 2928
ADR: 6.1,II
Merck Index: 14,3822

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR