Silver ≥99.9% (metals basis), casting grain 1.6-6.35 mm (0.063-0.25 in)

Supplier: Thermo Scientific Chemicals

40511-22 40511-30
CAAA40511-22EA 889.54 CAD
CAAA40511-22 CAAA40511-30
Silver ≥99.9% (metals basis), casting grain 1.6-6.35 mm (0.063-0.25 in)
Silver

Applications: Jewelry, alloys, electrical contacts


Purity: 99.9% CAS No.: 7440-22-4 Odor: Characteristic MDL No.: MFCD00003397


Jewelry, alloys, electrical contacts

Formula: Ag
Boiling Pt: 2212 °C (1013 hPa)
Melting Pt: 962 °C
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR